Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rangrf
Entrez Gene ID
100760667
Gene Name
RAN guanine nucleotide release factor
Gene Aliases
I79_002707
Chromosome Location
Chr.Un|NW_003613643: 1058557 - 1060207 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497172.2 XP_003497220.1 2 - 3 586 87
XM_007608217.1 XP_007606407.1 1 - 2 681 87
XM_007608221.1 XP_007606411.1 1 - 2 679 87
XM_007608224.1 XP_007606414.1 1 - 2 687 87

View Full Product Details