Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Yipf7
Entrez Gene ID
100752198
Gene Name
Yip1 domain family, member 7
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616599: 105716 - 112170 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514433.2 XP_003514481.1 1 - 2 144 105

View Full Product Details