Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdcd1
Entrez Gene ID
100774461
Gene Name
programmed cell death 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613726: 1453922 - 1466383 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499266.1 XP_003499314.1 3 - 4 599 64

View Full Product Details