Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spsb1
Entrez Gene ID
100768949
Gene Name
splA/ryanodine receptor domain and SOCS box containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615008: 245161 - 288300 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511018.2 XP_003511066.1 2 - 3 942 99
XM_007651582.1 XP_007649772.1 2 - 3 866 99

View Full Product Details