Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd50
Entrez Gene ID
100756565
Gene Name
ankyrin repeat domain 50
Gene Aliases
I79_016583
Chromosome Location
Chr.Un|NW_003614564: 350405 - 381348 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508598.2 XP_003508646.1 3 - 4 1531 78
XM_007649239.1 XP_007647429.1 3 - 4 1251 78

View Full Product Details