Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zscan10
Entrez Gene ID
100773821
Gene Name
zinc finger and SCAN domain containing 10
Gene Aliases
I79_016721
Chromosome Location
Chr.Un|NW_003614574: 551585 - 558440 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649332.1 XP_007647522.1 5 - 6 692 97

View Full Product Details