Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spaca3
Entrez Gene ID
100762682
Gene Name
sperm acrosome associated 3
Gene Aliases
I79_001068
Chromosome Location
Chr.Un|NW_003613601: 2533003 - 2537091 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495788.1 XP_003495836.1 1 - 2 185 67

View Full Product Details