Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scd5
Entrez Gene ID
100768175
Gene Name
stearoyl-CoA desaturase 5
Gene Aliases
I79_019372
Chromosome Location
Chr.Un|NW_003614991: 317414 - 355476 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651525.1 XP_007649715.1 1 - 2 253 88

View Full Product Details