Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Elane
Entrez Gene ID
100752732
Gene Name
elastase, neutrophil expressed
Gene Aliases
I79_009202
Chromosome Location
Chr.Un|NW_003613904: 661957 - 663846 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502604.1 XP_003502652.1 1 - 2 64 57

View Full Product Details