Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx20
Entrez Gene ID
100757776
Gene Name
sorting nexin 20
Gene Aliases
I79_005605
Chromosome Location
Chr.Un|NW_003613741: 659967 - 667252 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007640277.1 XP_007638467.1 3 - 4 399 69

View Full Product Details