Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Inhbe
Entrez Gene ID
100756167
Gene Name
inhibin, beta E
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614393: 418008 - 420121 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507497.2 XP_003507545.1 1 - 2 571 63

View Full Product Details