Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vamp1
Entrez Gene ID
100752483
Gene Name
vesicle-associated membrane protein 1 (synaptobrevin 1)
Gene Aliases
I79_001386
Chromosome Location
Chr.Un|NW_003613610: 3522445 - 3529325 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007646545.1 XP_007644735.1 4 - 5 495 73

View Full Product Details