Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc29
Entrez Gene ID
100760321
Gene Name
leucine rich repeat containing 29
Gene Aliases
I79_016518
Chromosome Location
Chr.Un|NW_003614552: 245676 - 260543 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508543.2 XP_003508591.2 7 - 8 1900 79

View Full Product Details