Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MANEAL
Entrez Gene ID
102173525
Gene Name
mannosidase endo-alpha like
Gene Aliases
-
Chromosome Location
Chr.3: 12685595 - 12694129 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005678678.3 XP_005678735.3 2 - 3 740 70
GenBank mRNA JO638869.2 - 2 - 3 1818 70

View Full Product Details