The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_018054445.1 | XP_017909934.1 | 2 - 3 | 1608 | 55 |
XM_018054447.1 | XP_017909936.1 | 2 - 3 | 1608 | 55 |