Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLX2
Entrez Gene ID
102182234
Gene Name
T cell leukemia homeobox 2
Gene Aliases
-
Chromosome Location
Chr.11: 9966389 - 9968595 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018054992.1 XP_017910481.1 1 - 2 739 85

View Full Product Details