Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELF5
Entrez Gene ID
102169781
Gene Name
E74 like ETS transcription factor 5
Gene Aliases
-
Chromosome Location
Chr.15: 17861189 - 17911965 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001314285.1 NP_001301214.1 4 - 5 451 106
XM_005690078.3 XP_005690135.1 4 - 5 813 106
XM_005690080.3 XP_005690137.1 4 - 5 540 106
GenBank mRNA KA331617.1 - 3 - 4 439 106
KF669900.1 - 4 - 5 451 106

View Full Product Details