Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD50
Entrez Gene ID
102177082
Gene Name
ankyrin repeat domain 50
Gene Aliases
-
Chromosome Location
Chr.17: 37955552 - 37993074 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018061515.1 XP_017917004.1 4 - 5 5064 105

View Full Product Details