Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DHRS7B
Entrez Gene ID
102173412
Gene Name
dehydrogenase/reductase 7B
Gene Aliases
-
Chromosome Location
Chr.19: 34841338 - 34862384 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693629.3 XP_005693686.2 2 - 3 317 78
XM_013972217.2 XP_013827671.1 3 - 4 485 78
XM_013972218.2 XP_013827672.1 2 - 3 349 78
GenBank mRNA JO589125.2 - 3 - 4 1015 78

View Full Product Details