Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC25A30
Entrez Gene ID
102179959
Gene Name
solute carrier family 25 member 30
Gene Aliases
-
Chromosome Location
Chr.12: 70992515 - 71016924 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005687432.3 XP_005687489.1 7 - 8 1201 88
XM_018056765.1 XP_017912254.1 7 - 8 785 88

View Full Product Details