Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC69
Entrez Gene ID
102190802
Gene Name
leucine rich repeat containing 69
Gene Aliases
-
Chromosome Location
Chr.14: 8913490 - 8989514 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005689249.3 XP_005689306.2 4 - 5 581 131

View Full Product Details