Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VWA2
Entrez Gene ID
102188112
Gene Name
von Willebrand factor A domain containing 2
Gene Aliases
-
Chromosome Location
Chr.26: 17230084 - 17281546 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018041456.1 XP_017896945.1 6 - 7 995 86
XM_018041457.1 XP_017896946.1 6 - 7 995 86

View Full Product Details