Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GADD45B
Entrez Gene ID
102183684
Gene Name
growth arrest and DNA damage inducible beta
Gene Aliases
-
Chromosome Location
Chr.7: 89446314 - 89448452 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018050778.1 XP_017906267.1 2 - 3 381 82
GenBank mRNA JO406036.2 - 2 - 3 557 82

View Full Product Details