Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB16
Entrez Gene ID
102186799
Gene Name
ankyrin repeat and SOCS box containing 16
Gene Aliases
-
Chromosome Location
Chr.19: 43626654 - 43634560 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018065210.1 XP_017920699.1 4 - 5 1272 87

View Full Product Details