Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MB21D2
Entrez Gene ID
102177752
Gene Name
Mab-21 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 74237472 - 74367072 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005675249.3 XP_005675306.2 1 - 2 770 97
GenBank mRNA JT717233.1 - 1 - 2 195 97
KA341724.1 - 1 - 2 392 97

View Full Product Details