Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLT6D1
Entrez Gene ID
102181049
Gene Name
glycosyltransferase 6 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.11: 102736917 - 102750983 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005687296.3 XP_005687353.2 4 - 5 353 105

View Full Product Details