The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173061.1 | NP_001166532.1 | 26 - 27 | 3778 | 80 |
GenBank mRNA | D84339.1 | - | 26 - 27 | 3778 | 80 |