Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Nppa
- Entrez Gene ID
-
100135578
- Gene Name
- natriuretic peptide A
- Gene Aliases
-
-
- Chromosome Location
-
Chr.Un|NT_176394: 22517564 - 22519052 on Build Cavpor3.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
X58562.1 |
- |
1 - 2 |
46 |
70 |
View Full Product Details