Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spt-I
Entrez Gene ID
36448
Gene Name
Serine palmitoyltransferase subunit I
Gene Aliases
CG4016, Dmel\CG4016, Dmel_CG4016, SPT, SPT-I, SPT1, Spt1, dSpt1, l(2)49Fb, l(2)SH1626, l(2)SH2 1626, lcb1
Chromosome Location
Chr.2R: 13168498 - 13170744 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086532
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136998.2 NP_610842.1 4 - 5 1304 111
NM_165967.4 NP_725255.1 5 - 6 1376 111
NM_165968.4 NP_725256.1 5 - 6 1346 111
GenBank mRNA BT010234.1 - 5 - 6 1344 111

View Full Product Details