Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sano
Entrez Gene ID
37185
Gene Name
serrano
Gene Aliases
CG12758, Dmel\CG12758, Dmel_CG12758
Chromosome Location
Chr.2R: 18878391 - 18959500 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034408
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144234.2 NP_001137706.1 4 - 5 1757 73
NM_001299666.1 NP_001286595.1 2 - 3 1422 73
NM_079064.4 NP_523788.2 2 - 3 1422 73
NM_166314.3 NP_725838.1 4 - 5 1501 73
NM_166315.3 NP_725839.1 5 - 6 1804 73
NM_166316.3 NP_725840.1 2 - 3 1656 73
GenBank mRNA BT012302.1 - 5 - 6 1801 73

View Full Product Details