Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acyp
Entrez Gene ID
34807
Gene Name
Acylphosphatase
Gene Aliases
ACPDro, AcPDro, BG:DS00180.1, CG16870, Dmel\CG16870, Dmel_CG16870
Chromosome Location
Chr.2L: 13912083 - 13912652 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025115
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298987.1 NP_001285916.1 1 - 1 428 88
NM_078839.3 NP_523563.1 1 - 1 428 88
GenBank mRNA AJ243543.1 - 1 - 1 310 88
KX532039.1 - 1 - 1 310 88

View Full Product Details