Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cul4
- Entrez Gene ID
-
35780
- Gene Name
- Cullin 4
- Gene Aliases
-
CG8711, CUL4, Cul-4, Cullin4, DMcul-4, Dmel\CG8711, Dmel_CG8711, cul-4, cul4, dCul4
- Chromosome Location
-
Chr.2R: 8102260 - 8106318 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033260
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136508.4 |
NP_610352.2 |
11 - 12 |
3061 |
71 |
GenBank mRNA |
BT050475.1 |
- |
11 - 12 |
3008 |
71 |
View Full Product Details