Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zip
Entrez Gene ID
38001
Gene Name
zipper
Gene Aliases
CG15792, DROMHC, Dm nmII, Dmel\CG15792, Dmel_CG15792, DmnmII, Dronm-MII, E(br), MHC, Mhc-c, Mhc-c[1], Myo, Myo II, Myo-II, Myo-II HC, Myo15, Myo2, MyoII, NMM, NMMII, ZIP, Zip, anon-WO0140519.37, l(2)02957, l(2)17F1, myo II, myoII, myosin-II, nmMHC, nmy-2, zip/MyoII, zpr
Chromosome Location
Chr.2R: 24990570 - 25011965 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0265434
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014552.3 NP_001014552.1 12 - 13 6039 78
NM_001014553.3 NP_001014553.1 12 - 13 5975 78
NM_001202079.1 NP_001189008.1 12 - 13 6002 78
NM_001202080.2 NP_001189009.1 13 - 14 5999 78
NM_001202081.1 NP_001189010.1 12 - 13 5996 78
NM_001259587.2 NP_001246516.1 13 - 14 6063 78
NM_079136.4 NP_523860.2 13 - 14 6159 78
NM_166704.4 NP_726506.1 13 - 14 6095 78

View Full Product Details