Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
klar
Entrez Gene ID
38067
Gene Name
klarsicht
Gene Aliases
CG17046, Dmel\CG17046, Dmel_CG17046, Klar, kls, marb
Chromosome Location
Chr.3L: 434083 - 540614 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001316
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043111.3 NP_001036576.1 10 - 11 3718 126
NM_001103993.2 NP_001097463.1 11 - 12 4369 126
NM_001274282.2 NP_001261211.1 10 - 11 4275 126
NM_001274283.2 NP_001261212.1 7 - 8 3191 126
NM_001274284.1 NP_001261213.1 2 - 3 597 126
NM_001299959.1 NP_001286888.1 7 - 8 3191 126
NM_079149.5 NP_523873.2 11 - 12 4369 126
GenBank mRNA BT010042.1 - 10 - 11 3308 126

View Full Product Details