Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cbp80
Entrez Gene ID
44409
Gene Name
cap binding protein 80
Gene Aliases
CBP80, CG7035, Dmel\CG7035, Dmel_CG7035, EG:84H4.3, cbp80
Chromosome Location
Chr.X: 4650015 - 4658259 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0022942
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080011.3 NP_524750.2 3 - 4 1762 68
NM_167012.3 NP_726938.1 3 - 4 1797 68
GenBank mRNA AY051796.1 - 3 - 4 1737 68

View Full Product Details