Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GstE2
Entrez Gene ID
37107
Gene Name
Glutathione S transferase E2
Gene Aliases
CG17523, DmGSTE2, Dmel\CG17523, Dmel_CG17523, GSTE2, gste2
Chromosome Location
Chr.2R: 18399486 - 18400375 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0063498
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137480.4 NP_611324.1 1 - 1 650 82
GenBank mRNA AY071644.1 - 1 - 1 646 82

View Full Product Details