Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- eEFSec
- Entrez Gene ID
-
37444
- Gene Name
- eukaryotic translation elongation factor, selenocysteine-specific
- Gene Aliases
-
CG9841, Dmel\CG9841, Dmel_CG9841, EFSec, EFsec, EfSec, Efsec, Eftu, SelB/eEFsec, dEFsec, deEFsec, eEFsec
- Chromosome Location
-
Chr.2R: 21384552 - 21386221 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034627
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137740.3 |
NP_611584.1 |
1 - 2 |
331 |
67 |
GenBank mRNA |
AY071627.1 |
- |
1 - 2 |
325 |
67 |
View Full Product Details