Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mud
Entrez Gene ID
44839
Gene Name
mushroom body defect
Gene Aliases
CG12047, Dmel\CG12047, Dmel_CG12047, KS63, Mud, NuMA, NuMa
Chromosome Location
Chr.X: 14247732 - 14258595 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0002873
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258736.2 NP_001245665.1 10 - 11 7041 70
NM_001272616.1 NP_001259545.1 11 - 12 6657 70

View Full Product Details