Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mud
Entrez Gene ID
44839
Gene Name
mushroom body defect
Gene Aliases
CG12047, Dmel\CG12047, Dmel_CG12047, KS63, Mud, NuMA, NuMa
Chromosome Location
Chr.X: 14247732 - 14258595 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0002873
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258736.2 NP_001245665.1 5 - 6 1406 77
NM_001258737.2 NP_001245666.1 5 - 6 1406 77
NM_001272615.2 NP_001259544.1 5 - 6 1406 77
NM_001272616.1 NP_001259545.1 5 - 6 1406 77
NM_080495.6 NP_536743.3 5 - 6 1406 77
NM_167399.6 NP_727769.3 5 - 6 1406 77
NM_167400.5 NP_727770.2 5 - 6 1406 77

View Full Product Details