Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mthl14
Entrez Gene ID
318046
Gene Name
methuselah-like 14
Gene Aliases
BcDNA:RE31310, CG32476, Dmel\CG32476, Dmel_CG32476
Chromosome Location
Chr.3L: 247491 - 250305 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0052476
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_167821.3 NP_728509.1 3 - 4 683 65
GenBank mRNA AY071287.1 - 3 - 4 679 65

View Full Product Details