Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp6d2
Entrez Gene ID
37594
Gene Name
CG4373-PA
Gene Aliases
6d2, CG4373, Dmel\CG4373, Dmel_CG4373
Chromosome Location
Chr.2R: 22652573 - 22654530 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034756
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137854.3 NP_611698.1 3 - 4 1488 67
GenBank mRNA BT009998.1 - 3 - 4 1485 67

View Full Product Details