Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rpp30
Entrez Gene ID
44392
Gene Name
RNaseP protein p30
Gene Aliases
#19/1, CG11606, Dmel\CG11606, Dmel_CG11606, RNase P, Rpp1, coconut, l(2)k01901
Chromosome Location
Chr.2L: 251039 - 252373 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0283652
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272879.1 NP_001259808.1 4 - 5 673 67
NM_080010.4 NP_524749.1 4 - 5 673 67
GenBank mRNA AY075315.1 - 4 - 5 619 67

View Full Product Details