Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZC3H3
Entrez Gene ID
38968
Gene Name
CG6694-PA
Gene Aliases
CG6694, Dmel\CG6694, Dmel_CG6694, dZC3H3
Chromosome Location
Chr.3L: 8507469 - 8509763 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035900
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139973.3 NP_648230.1 3 - 4 1531 68
GenBank mRNA BT010061.1 - 3 - 4 1516 68

View Full Product Details