Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GluRIA
- Entrez Gene ID
-
38742
- Gene Name
- Glutamate receptor IA
- Gene Aliases
-
CG8442, DGluR-I, DGluR-IA, DGluRI, DmelGlu-R1, Dmel\CG8442, Dmel_CG8442, Dro GluRIA, Glu-R1, Glu-RI, GluR1, GluR1A, GluRI, dGluRI, dglur-IA
- Chromosome Location
-
Chr.3L: 6624032 - 6635059 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004619
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057507.4 |
NP_476855.1 |
2 - 3 |
819 |
80 |
GenBank mRNA |
BT050435.1 |
- |
2 - 3 |
817 |
80 |
View Full Product Details