Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRMP
Entrez Gene ID
40675
Gene Name
Collapsin Response Mediator Protein
Gene Aliases
BcDNA:HL02693, CG1411, DHP, Dmel\CG1411, Dmel_CG1411, EP(3)3238, EP3238, Pyd2, crmp, l(3)83Ba, pyd2
Chromosome Location
Chr.3R: 5571108 - 5579189 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0023023
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057959.3 NP_477307.1 4 - 5 463 109
NM_169070.2 NP_730952.1 4 - 5 463 109
NM_169072.3 NP_730954.2 4 - 5 463 109
GenBank mRNA AF077837.1 - 4 - 5 449 109
AF160900.1 - 4 - 5 442 109
AF465756.1 - 3 - 4 330 109
GU452673.1 - 4 - 5 418 109
GU452674.1 - 4 - 5 415 109
GU452675.1 - 4 - 5 423 109

View Full Product Details