Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rga
Entrez Gene ID
40683
Gene Name
Regena
Gene Aliases
CG2138, CG2161, Dmel\CG2161, Dmel_CG2161, NOT2, Not2, RGA, l(3)03834, not2, rga
Chromosome Location
Chr.3R: 5607275 - 5613112 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0017550
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300254.1 NP_001287183.1 3 - 4 1091 64
NM_079515.3 NP_524239.2 3 - 4 1393 64
NM_169080.3 NP_730966.1 3 - 4 1393 64
GenBank mRNA BT025208.2 - 3 - 4 1365 64

View Full Product Details