Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Calr
Entrez Gene ID
41166
Gene Name
Calreticulin
Gene Aliases
CG9429, Clr, Crc, Dmel\CG9429, Dmel_CG9429, anon-EST:Liang-2.11, anon-EST:Liang-2.17, anon-EST:fe3D7, chr3R:5454619..5454778, clone 2.11, clone 2.17, crc, eth, potp
Chromosome Location
Chr.3R: 9628691 - 9630851 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005585
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275501.1 NP_001262430.1 2 - 3 282 68
NM_079569.5 NP_524293.2 2 - 3 282 68
GenBank mRNA BT001670.1 - 2 - 3 274 68

View Full Product Details