Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTA1-like
Entrez Gene ID
40693
Gene Name
Metastasis associated 1-like
Gene Aliases
CG2244, Dmel\CG2244, Dmel_CG2244, MTA, MTA-like, MTA1, dMTA, dMTA-1, dMTA1-like, p120
Chromosome Location
Chr.3R: 5638655 - 5648738 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027951
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144539.2 NP_001138011.1 7 - 8 2463 69
NM_001275364.2 NP_001262293.1 9 - 10 2118 69
NM_141309.4 NP_649566.2 8 - 9 2010 69
NM_169086.3 NP_730976.1 9 - 10 2118 69

View Full Product Details