Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Usp1
Entrez Gene ID
43462
Gene Name
Ubiquitin specific protease 1
Gene Aliases
BcDNA:LD22910, CG15817, Dmel\CG15817, Dmel_CG15817, LD22910, Q8I077, USP1, usp1
Chromosome Location
Chr.3R: 29229161 - 29235302 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028476
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276132.1 NP_001263061.1 5 - 6 2404 76
NM_143430.4 NP_651687.3 4 - 5 2564 76
NM_170403.3 NP_733282.1 5 - 6 2404 76
GenBank mRNA AF181650.1 - 4 - 5 2542 76
BT001496.1 - 5 - 6 2377 76

View Full Product Details