Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
woc
Entrez Gene ID
47249
Gene Name
without children
Gene Aliases
CG5965, Dmel\CG5965, Dmel_CG5965, WOC, Woc
Chromosome Location
Chr.3R: 27256595 - 27264055 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010328
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104476.2 NP_001097946.1 5 - 6 3339 65
NM_001276080.1 NP_001263009.1 4 - 5 3333 65
NM_001276081.1 NP_001263010.1 5 - 6 3336 65
NM_080147.3 NP_524886.2 4 - 5 3336 65

View Full Product Details